Learn More
BIOHIPPO Rat S100A3 Microsample
SDP
Catalog No. 5000314084
Encompass
Rat S100A3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A3 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 069 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat S100A3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat S100A3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat S100A3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14084 Supplier BIOHIPPO Supplier No. BHE15212394
Rat S100A3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A3 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 069 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat S100A3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat S100A3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat S100A3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color