Learn More
Biomatik Corporation Rat Secreted Frizzled Related Protein 5 (SFRP5) ELISA Kit
Supplier:  Biomatik Corporation EKU07224
Encompass_Preferred
Rat Secreted Frizzled Related Protein 5 (SFRP5) ELISA Kit for detecting SFRP5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 47pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZTV6. Gene ID: 309377. Target Synonyms: SARP3; Secreted Apoptosis Related Protein 3; Frizzled-related protein 1b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8770