Learn More
American Research Products Inc Rat secreted phospholipase A2
Catalog No. 5000329811
Encompass
Rat secreted phospholipase A2 sPLA2 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29811 Supplier American Research Products Inc Supplier No. CSBE13206R
Rat secreted phospholipase A2 sPLA2 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich