Learn More
Biomatik Corporation Rat Secretin (SCT) ELISA Kit
Supplier:  Biomatik Corporation EKU11579
Encompass_Preferred
Rat Secretin (SCT) ELISA Kit for detecting SCT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.24pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P11384. Gene ID: 24769. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5637