Learn More
Biomatik Corporation Rat Secretory Leukocyte Peptidase Inhibitor (SLPI) ELISA Kit
Supplier:  Biomatik Corporation EKU11695
Rat Secretory Leukocyte Peptidase Inhibitor (SLPI) ELISA Kit for detecting SLPI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WUQ4. Gene ID: 84386. Target Synonyms: ALK1; ALP; MPI; BLPI; HUSI; HUSI-I; WAP4; WFDC4; Antileukoproteinase; Seminal proteinase inhibitor; Mucus proteinase inhibitor; WAP four-disulfide core domain protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8098