Learn More
Sino Biological Anti-E-Selectin/CD62E Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80033MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat E-Selectin / CD62e / SELE (rR E-Selectin / CD62e / SELE; Catalog#80033-R08H; P98105-1; Met 1-Pro 494). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9065