Learn More
Sino Biological Recombinant Anti-E-Selectin/CD62E Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80033R10220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat E-Selectin / CD62e / SELE (rR E-Selectin / CD62e / SELE; Catalog#80033-R08H; P98105-1; Met1-Pro494).
Catalog No. 50-241-1428