Learn More
Sino Biological Anti-E-Selectin/CD62E Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80033RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat E-Selectin / CD62e / SELE (rR E-Selectin / CD62e / SELE; Catalog#80033-R08H; P98105-1; Met 1-Pro 494). E-Selectin / CD62e / SELE specific IgG was purified by Rat E-Selectin / CD62e / SELE affinity chromatography.
Catalog No. 50-240-9031