Learn More
Sino Biological Rat SELE PAb 100ul (Sino Biological)
Supplier:  Sino Biological 80033RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat E-Selectin / CD62e / SELE (Catalog#80033-R08H; P98105-1; Met1-Pro494). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2217