Learn More
Biomatik Corporation Rat Selectin, Leukocyte (SELL) CLIA Kit
Supplier:  Biomatik Corporation EKU08816
Encompass_Preferred
Rat Selectin, Leukocyte (SELL) CLIA Kit for detecting SELL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 41.2-30,000pg/ml. Sensitivity: 17.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P30836. Target Synonyms: CD62L; L-Selectin; SEL-L; LAM1; LECAM1; LNHR; LSEL; LYAM1; Leu-8; Lyam-1; PLNHR; TQ1; HLHRc; MEL14; Lymphocyte Adhesion Molecule 1; Lymph Node Homing Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6801