Learn More
Biomatik Corporation Rat Selectin, Leukocyte (SELL) ELISA Kit
Supplier:  Biomatik Corporation EKU07248
Encompass_Preferred
Rat Selectin, Leukocyte (SELL) ELISA Kit for detecting SELL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30836. Target Synonyms: CD62L; L-Selectin; SEL-L; LAM1; LECAM1; LNHR; LSEL; LYAM1; Leu-8; Lyam-1; PLNHR; TQ1; HLHRc; MEL14; Lymphocyte Adhesion Molecule 1; Lymph Node Homing Receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4728