Learn More
American Research Products Inc Rat selenoprotein P plasma 1
Catalog No. 5000327748
Encompass
Rat selenoprotein P plasma 1 (SEPP1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27748 Supplier American Research Products Inc Supplier No. CSBE17683R
Rat selenoprotein P plasma 1 (SEPP1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich