Learn More
BIOHIPPO Rat SELP Microsample
SDP
Catalog No. 5000315193
Encompass
Rat SELP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SELP in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 72 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SELP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SELP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SELP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15193 Supplier BIOHIPPO Supplier No. BHE15212614
Rat SELP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SELP in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 72 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SELP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SELP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SELP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color