Learn More
Sino Biological Rat SELP PAb 100ul (Sino Biological)
Supplier:  Sino Biological 80451T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat SELP / P Selectin (Catalog#80451-R08H; AAI01920.1; Met1-Gln700). SELP / P Selectin specific IgG was purified by Rat SELP / P Selectin affinity chromatography.
Catalog No. 50-237-0802