Learn More
Sino Biological Rat SEMA4D Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80320R02H100
Encompass_Preferred
A DNA sequence encoding the rat SEMA4D (NP_001164034.1)(Met1-Met712) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2132