Learn More
American Research Products Inc Rat Serine threonine protein k
Catalog No. 5000328263
Encompass
Rat Serine/threonine-protein kinase TNNI3K (TNNI3K) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 69 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28263 Supplier American Research Products Inc Supplier No. CSBEL024014RA
Rat Serine/threonine-protein kinase TNNI3K (TNNI3K) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 69 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich