Learn More
Biomatik Corporation Rat Serine/threonine-protein kinase mTOR (mTOR) ELISA Kit
Supplier:  Biomatik Corporation EKU09086
Encompass_Preferred
Rat Serine/threonine-protein kinase mTOR (mTOR) ELISA Kit for detecting mTOR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42346. Gene ID: 56718. Target Synonyms: FRAP; FRAP1; FRAP2; MTOR; RAFT1; RAPT1; FK506 Binding Protein 12 Rapamycin Associated Protein; Mammalian Target Of Rapamycin; FKBP12-Rapamycin Complex-Associated Protein 1; Rapamycin and FKBP12 target. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5476