Learn More
American Research Products Inc Rat Serotonin Receptor 2A 5 hy
Catalog No. 5000328702
Encompass
Rat Serotonin Receptor 2A/5-hydroXytryptamine Receptor 2A (5-HT-2A) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28702 Supplier American Research Products Inc Supplier No. CSBE14956R
Rat Serotonin Receptor 2A/5-hydroXytryptamine Receptor 2A (5-HT-2A) ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich