Learn More
Sino Biological Recombinant Anti-Serpin A1/Alpha-1-antitrypsin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80476R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat SerpinA1 (rR SerpinA1; Catalog#80476-R08H; NP_071964.2; Met1-Arg411).
Catalog No. 50-240-8905