Learn More
Sino Biological Anti-Serpin A1/Alpha-1-antitrypsin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80476T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat SerpinA1 (rR SerpinA1; Catalog#80476-R08H; NP_071964.2; Met1-Arg411). SerpinA1 specific IgG was purified by Rat SerpinA1 affinity chromatography.
Catalog No. 50-240-8496