Learn More
Cusabio Technology LLC RAT SERPINA12 ELISA KIT
Catalog No. 5000049240
Encompass_Preferred
This kit detects SERPINA12 (vaspin) in rat samples including serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 31 25 pg/mL to 2000 pg/mL with a sensitivity limit of 7 8 pg/mL providing precise measurement of visceral adipose-specific serine protease inhibitor levels The assay requires 50-100 L of sample per test and can be completed within 1-5 hours with absorbance readings taken at 450 nm Designed for research applications this reliable kit enables the study of metabolic regulation and adipose tissue function supporting investigations in obesity insulin resistance and related metabolic disorders through accurate quantification of SERPINA12 protein concentrations
Catalog No. 50-000-49240 Supplier Cusabio Technology LLC Supplier No. CSBE09813R
This kit detects SERPINA12 (vaspin) in rat samples including serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 31 25 pg/mL to 2000 pg/mL with a sensitivity limit of 7 8 pg/mL providing precise measurement of visceral adipose-specific serine protease inhibitor levels The assay requires 50-100 L of sample per test and can be completed within 1-5 hours with absorbance readings taken at 450 nm Designed for research applications this reliable kit enables the study of metabolic regulation and adipose tissue function supporting investigations in obesity insulin resistance and related metabolic disorders through accurate quantification of SERPINA12 protein concentrations