Learn More
Sino Biological Recombinant Anti-PAI-1/Serpin E1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80442R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat PAI-1 / SerpinE1 (rR PAI-1 / SerpinE1; Catalog#80442-R08H; NP_036752.1; Met1-Pro402).
Catalog No. 50-240-8179