Learn More
Sino Biological Anti-PAI-1/Serpin E1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80442RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat PAI-1 / SerpinE1 (rR PAI-1 / SerpinE1; Catalog#80442-R08H; NP_036752.1; Met1-Pro402). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8379