Learn More
Sino Biological Rat SerpinE1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 14244MM01100
Encompass_Preferred
A DNA sequence encoding the rat SERPINE1 (NP_036752.1) (Met1-Pro402) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4046