Learn More
Sino Biological Rat SerpinF2 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 80687R08H50
Encompass_Preferred
A DNA sequence encoding the rat SERPINF2 (NP_001011892.1) (Met1-Lys491) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2852