Learn More
Rockland Immunochemicals Rat Serum Non-Sterile 50 mL
SDP
Catalog No. 501943966
Encompass_Preferred
Rat Serum is used as a component of bioassays immunoassays or enzyme assays. Rat Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Rat serum is also available as a sterile preparation for use as a cell culture supplement.
Catalog No. 50-194-3966 Supplier Rockland Immunochemicals Supplier No. D210000050
Rat Serum is used as a component of bioassays immunoassays or enzyme assays. Rat Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Rat serum is also available as a sterile preparation for use as a cell culture supplement.