Learn More
American Research Products Inc Rat seX hormone binding globul
Catalog No. 5000330240
Encompass
Rat seX hormone-binding globulin (SHBG) ELISA kit from CUSABIO Detection Range 375 ng/mL-6000 ng/mL Sensitivity 375 ng/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Competitive
Catalog No. 50-003-30240 Supplier American Research Products Inc Supplier No. CSBE12118R
Rat seX hormone-binding globulin (SHBG) ELISA kit from CUSABIO Detection Range 375 ng/mL-6000 ng/mL Sensitivity 375 ng/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Competitive