Learn More
BIOHIPPO Rat SHH Microsample
SDP
Catalog No. 5000314071
Encompass
Rat SHH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SHH in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 55 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SHH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SHH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SHH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14071 Supplier BIOHIPPO Supplier No. BHE15211142
Rat SHH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SHH in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 55 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SHH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SHH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SHH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color