Learn More
Biomatik Corporation Rat Sialidase 2, Cytosolic (SIAL2) ELISA Kit
Supplier:  Biomatik Corporation EKU07330
Encompass_Preferred
Rat Sialidase 2, Cytosolic (SIAL2) ELISA Kit for detecting SIAL2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64627. Target Synonyms: MEU2; N-Acetyl-Alpha-Neuraminidase 2; Cytosolic sialidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8758