Learn More
Biomatik Corporation Rat Sirtuin 1 (SIRT1) CLIA Kit
Supplier:  Biomatik Corporation EKU09975
Encompass_Preferred
Rat Sirtuin 1 (SIRT1) CLIA Kit for detecting SIRT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: A0A8I6A306. Target Synonyms: SIR2L1; Silent Mating Type Information Regulation 2 Homolog 1; NAD-Dependent Deacetylase Sirtuin-1; SIR2-like protein 1; SirtT1 75 kDa fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6894