Learn More
Biomatik Corporation Rat Sirtuin 7 (SIRT7) ELISA Kit
Supplier:  Biomatik Corporation EKU11429
Encompass_Preferred
Rat Sirtuin 7 (SIRT7) ELISA Kit for detecting SIRT7 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B2RZ55. Gene ID: 303745. Target Synonyms: SIR2L7; NAD-Dependent Deacetylase Sirtuin-7; Silent Mating Type Information Regulation 2 Homolog 7; Regulatory protein SIR2 homolog 7; SIR2-like protein 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8144