Learn More
Innovative Research Inc Rat Skel Mscl Tssu Lyst 100ug
Catalog No. 502743996
Encompass_Preferred
Rat Skeletal Muscle Tissue Lysate 100ug - Rat Skeletal Muscle Tissue Lysate from Innovative Research is provided as a Liquid buffered in SDS sample buffer containing 5% beta-mercaptoethanol. This product is especially useful for applications like Western Blot, among others. For continuous use, store at 2-8–‘C. If not using the entire volume immediately, store single-use aliquots at -70–‘C. Repeated freeze/thaw cycles are not recommended. Under proper storage conditions, the shelf life is six months from the date of receipt. Additional Details Species Rat Source Skeletal Muscle tissue lysate Format Liquid Storage Conditions -70C.
Catalog No. 50-274-3996 Supplier Innovative Research Inc Supplier No. IRTSKMTL100UG
Rat Skeletal Muscle Tissue Lysate 100ug - Rat Skeletal Muscle Tissue Lysate from Innovative Research is provided as a Liquid buffered in SDS sample buffer containing 5% beta-mercaptoethanol. This product is especially useful for applications like Western Blot, among others. For continuous use, store at 2-8–‘C. If not using the entire volume immediately, store single-use aliquots at -70–‘C. Repeated freeze/thaw cycles are not recommended. Under proper storage conditions, the shelf life is six months from the date of receipt. Additional Details Species Rat Source Skeletal Muscle tissue lysate Format Liquid Storage Conditions -70C.