Learn More
Sino Biological Rat SLAMF1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80349R08H100
Encompass_Preferred
A DNA sequence encoding the rat SLAMF1 (D3ZAD7) (Met1-Leu242) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2925