Learn More
Sino Biological Anti-Band 3/SLC4A1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107891T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Rat SLC4A1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8402