Learn More
Biomatik Corporation Rat Smoothelin (SMTN) ELISA Kit
Supplier:  Biomatik Corporation EKU07379
Encompass_Preferred
Rat Smoothelin (SMTN) ELISA Kit for detecting SMTN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.25ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4ABA5. Gene ID: 289734. Target Synonyms: SMSMO. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7640