Learn More
BIOHIPPO Rat SMTN Microsample
SDP
Catalog No. 5000315656
Encompass
Rat SMTN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SMTN in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 25 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SMTN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SMTN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SMTN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15656 Supplier BIOHIPPO Supplier No. BHE15210939
Rat SMTN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SMTN in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 25 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SMTN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SMTN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SMTN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color