Learn More
BIOHIPPO Rat SOD1 Microsample
SDP
Catalog No. 5000315558
Encompass
Rat SOD1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SOD1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SOD1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SOD1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SOD1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15558 Supplier BIOHIPPO Supplier No. BHE15212520
Rat SOD1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SOD1 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat SOD1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat SOD1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat SOD1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color