Learn More
American Research Products Inc Rat Sodium potassium transport
Catalog No. 5000327421
Encompass
Rat Sodium/potassium-transporting ATPase subunit alpha-1 (ATP1A1) ELISA kit from CUSABIO Detection Range 31 2 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27421 Supplier American Research Products Inc Supplier No. CSBEL002322RA
Rat Sodium/potassium-transporting ATPase subunit alpha-1 (ATP1A1) ELISA kit from CUSABIO Detection Range 31 2 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich