Learn More
Biomatik Corporation Rat Sodium/Glucose Cotransporter 2 (SGLT2) ELISA Kit
Supplier:  Biomatik Corporation EKU07387
Encompass_Preferred
Rat Sodium/Glucose Cotransporter 2 (SGLT2) ELISA Kit for detecting SGLT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53792. Gene ID: 64522. Target Synonyms: SLC5A2; Solute Carrier Family 5 Member 2; Low affinity sodium-glucose cotransporter; Na (+) /glucose cotransporter 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8756