Learn More
Biomatik Corporation Rat Sodium/potassium Transporting ATPase Subunit Beta-3 (ATP1b3) ELISA Kit
Supplier:  Biomatik Corporation EKU02637
Encompass_Preferred
Rat Sodium/potassium Transporting ATPase Subunit Beta-3 (ATP1b3) ELISA Kit for detecting ATP1b3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63377. Gene ID: 25390. Target Synonyms: CD298; ATP1-B3; ATPB-3; ATPase, Na+/K+ Transporting Beta 3 Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6081