Learn More
American Research Products Inc Rat soluble Cluster of differe
Catalog No. 5000328956
Encompass
Rat soluble Cluster of differentiation 86 sCD86 ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28956 Supplier American Research Products Inc Supplier No. CSBE08543R
Rat soluble Cluster of differentiation 86 sCD86 ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich