Learn More
American Research Products Inc Rat Soluble Cluster of differe
Catalog No. 5000328959
Encompass
Rat Soluble Cluster of differentiation 40 ligand sCD40L ELISA Kit from CUSABIO Detection Range 1 87 ng/mL-120 ng/mL Sensitivity 0 47 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28959 Supplier American Research Products Inc Supplier No. CSBE07395R
Rat Soluble Cluster of differentiation 40 ligand sCD40L ELISA Kit from CUSABIO Detection Range 1 87 ng/mL-120 ng/mL Sensitivity 0 47 ng/mL Sample serum plasma tissue homogenates Method Sandwich