Learn More
American Research Products Inc Rat soluble fibrin monomer com
Catalog No. 5000330061
Encompass
Rat soluble fibrin monomer compleX (SFMC) ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 6 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30061 Supplier American Research Products Inc Supplier No. CSBE16270R
Rat soluble fibrin monomer compleX (SFMC) ELISA Kit from CUSABIO Detection Range 62 5 ng/mL-4000 ng/mL Sensitivity 15 6 ng/mL Sample serum plasma tissue homogenates Method Sandwich