Learn More
American Research Products Inc Rat Soluble Lectin like OXidiz
Catalog No. 5000328514
Encompass
Rat Soluble Lectin-like OXidized Low Density Lipoprotein Receptor-1 (sLOX-1) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28514 Supplier American Research Products Inc Supplier No. CSBE15845R
Rat Soluble Lectin-like OXidized Low Density Lipoprotein Receptor-1 (sLOX-1) ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich