Learn More
American Research Products Inc Rat soluble platelet endotheli
Catalog No. 5000329006
Encompass
Rat soluble platelet endothelial cell adhesion molecule 1 sPECAM-1 ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29006 Supplier American Research Products Inc Supplier No. CSBE08134R
Rat soluble platelet endothelial cell adhesion molecule 1 sPECAM-1 ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich