Learn More
American Research Products Inc Rat Soluble protein 100B S 10
Catalog No. 5000326434
Encompass
Rat Soluble protein-100B S-100B ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26434 Supplier American Research Products Inc Supplier No. CSBE08066R
Rat Soluble protein-100B S-100B ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich