Learn More
American Research Products Inc Rat soluble receptor actiVator
Catalog No. 5000329926
Encompass
Rat soluble receptor actiVator of nuclear factor-kB ligand sRANKL ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29926 Supplier American Research Products Inc Supplier No. CSBE05126R
Rat soluble receptor actiVator of nuclear factor-kB ligand sRANKL ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates Method Sandwich