Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat Somatostatin R1/SSTR1 (NP_036851) VersaClone cDNA
SDP

Catalog No. RDC1933
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC1933 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC1933 Supplier R&D Systems™ Supplier No. RDC1933

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Somatostatin Receptor Type 1

The Rat Somatostatin R1/SSTR1 (NP_036851) VersaClone cDNA clone contains the complete ORF for Somatostatin R1/SSTR1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Somatostatin R1/SSTR1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Product Type cDNA
Gene rSstr1
Accession Number NP_036851
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1189bp
Gene ID (Entrez) 6751 (Human); 20605 (Mouse); 25033 (Rat)
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Rat
Quantity 10 μg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.