Learn More
Biomatik Corporation Rat Sorbitol Dehydrogenase (SDH) ELISA Kit
Supplier:  Biomatik Corporation EKU07416
Encompass_Preferred
Rat Sorbitol Dehydrogenase (SDH) ELISA Kit for detecting SDH in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27867. Gene ID: 24788. Target Synonyms: SORD; L-iditol 2-dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6719