Learn More
Sino Biological Recombinant Anti-SOST/Sclerostin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80009R118100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Sclerostin/SOST (rR Sclerostin/SOST; Catalog#80009-R08H; NP_085073.1; Met1-Tyr213).
Catalog No. 50-255-2882